Assay Details
| Gene Symbol: Mansc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845870.3 | XP_004845927.1 | 3 - 4 | 657 | 67 | |
| XM_021259169.1 | XP_021114828.1 | 3 - 4 | 576 | 67 | ||
More Information
Set Membership: |
|

Mansc1
Genomic Map