Assay Details
| Gene Symbol: Mapre2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855208.3 | XP_004855265.1 | 4 - 5 | 783 | 69 | |
| XM_004855210.3 | XP_004855267.1 | 5 - 6 | 571 | 69 | ||
| XM_021236671.1 | XP_021092330.1 | 5 - 6 | 625 | 69 | ||
More Information
Set Membership: |
|

Mapre2
Genomic Map