Assay Details
| Gene Symbol: LOC101701428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857892.2 | XP_004857949.1 | 4 - 5 | 570 | 121 | |
| XM_021238724.1 | XP_021094383.1 | 3 - 4 | 519 | 121 | ||
More Information
Set Membership: |
|

LOC101701428
Genomic Map