Assay Details
| Gene Symbol: Ccdc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861021.3 | XP_004861078.1 | 12 - 13 | 2220 | 60 | |
| XM_004861024.3 | XP_004861081.1 | 12 - 13 | 2221 | 60 | ||
| XM_013078160.2 | XP_012933614.1 | 12 - 13 | 2222 | 60 | ||
| XM_021241001.1 | XP_021096660.1 | 12 - 13 | 2224 | 60 | ||
| XM_021241002.1 | XP_021096661.1 | 12 - 13 | 2225 | 60 | ||
More Information
Set Membership: |
|

Ccdc57
Genomic Map