Assay Details
Gene Symbol: Mia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013069057.2 | XP_012924511.1 | 2 - 3 | 348 | 91 | |
XM_021251733.1 | XP_021107392.1 | 3 - 4 | 352 | 91 | ||
XM_021251734.1 | XP_021107393.1 | 4 - 5 | 375 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
