Assay Details
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845375.2 | XP_004845432.1 | 5 - 6 | 615 | 58 | |
| XM_021258788.1 | XP_021114447.1 | 5 - 6 | 570 | 58 | ||
| XM_021258789.1 | XP_021114448.1 | 3 - 4 | 378 | 58 | ||
More Information
Set Membership: |
|

Cerk
Genomic Map