Assay Details
| Gene Symbol: Nkpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068625.2 | XP_012924079.2 | 2 - 3 | 1008 | 70 | |
| XM_021250868.1 | XP_021106527.1 | 2 - 3 | 1010 | 70 | ||
| XM_021250870.1 | XP_021106529.1 | 2 - 3 | 1017 | 70 | ||
| XM_021250871.1 | XP_021106530.1 | 2 - 3 | 1013 | 70 | ||
More Information
Set Membership: |
|

Nkpd1
Genomic Map