Assay Details
| Gene Symbol: E2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013074186.2 | XP_012929640.1 | 4 - 5 | 1199 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

E2f2| Gene Symbol: E2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013074186.2 | XP_012929640.1 | 4 - 5 | 1199 | 61 | |
Set Membership: |
|