Assay Details
| Gene Symbol: Ppm1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834209.2 | XP_004834266.1 | 9 - 10 | 1279 | 57 | |
| XM_004834210.2 | XP_004834267.1 | 12 - 13 | 1505 | 57 | ||
| XM_021241841.1 | XP_021097500.1 | 8 - 9 | 1171 | 57 | ||
| XM_021241920.1 | XP_021097579.1 | 8 - 9 | 1024 | 57 | ||
More Information
Set Membership: |
|

Ppm1m
Genomic Map