Assay Details
| Gene Symbol: Gfra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838862.2 | XP_004838919.1 | 6 - 7 | 1027 | 71 | |
| XM_004838865.2 | XP_004838922.1 | 5 - 6 | 1012 | 71 | ||
More Information
Set Membership: |
|

Gfra1
Genomic Map