Assay Details
| Gene Symbol: Sdcbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842149.3 | XP_004842206.1 | 5 - 6 | 571 | 70 | |
| XM_013071255.2 | XP_012926709.1 | 5 - 6 | 613 | 70 | ||
| XM_021256093.1 | XP_021111752.1 | 6 - 7 | 680 | 70 | ||
More Information
Set Membership: |
|

Sdcbp
Genomic Map