Assay Details
| Gene Symbol: Lca5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842361.2 | XP_004842418.1 | 5 - 6 | 1371 | 77 | |
| XM_013071318.2 | XP_012926772.1 | 4 - 5 | 1123 | 77 | ||
| XM_021256222.1 | XP_021111881.1 | 6 - 7 | 1579 | 77 | ||
| XM_021256223.1 | XP_021111882.1 | 6 - 7 | 1570 | 77 | ||
| XM_021256224.1 | XP_021111883.1 | 7 - 8 | 1629 | 77 | ||
| XM_021256225.1 | XP_021111884.1 | 5 - 6 | 1353 | 77 | ||
| XM_021256226.1 | XP_021111885.1 | 7 - 8 | 1598 | 77 | ||
| XM_021256227.1 | XP_021111886.1 | 6 - 7 | 1548 | 77 | ||
| XM_021256228.1 | XP_021111887.1 | 5 - 6 | 1668 | 77 | ||
| XM_021256229.1 | XP_021111888.1 | 5 - 6 | 1322 | 77 | ||
| XM_021256230.1 | XP_021111889.1 | 6 - 7 | 1559 | 77 | ||
| XM_021256231.1 | XP_021111890.1 | 5 - 6 | 1380 | 77 | ||
| XM_021256232.1 | XP_021111891.1 | 4 - 5 | 1154 | 77 | ||
| XM_021256233.1 | XP_021111892.1 | 4 - 5 | 1220 | 77 | ||
| XM_021256234.1 | XP_021111893.1 | 4 - 5 | 2335 | 77 | ||
| XM_021256235.1 | XP_021111894.1 | 4 - 5 | 1121 | 77 | ||
| XM_021256236.1 | XP_021111895.1 | 3 - 4 | 981 | 77 | ||
| XM_021256237.1 | XP_021111896.1 | 6 - 7 | 1579 | 77 | ||
| XM_021256238.1 | XP_021111897.1 | 6 - 7 | 1580 | 77 | ||
More Information
Set Membership: |
|

Lca5l
Genomic Map