Assay Details
| Gene Symbol: Mrpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854096.2 | XP_004854153.1 | 1 - 2 | 174 | 55 | |
| XM_021265953.1 | XP_021121612.1 | 1 - 2 | 174 | 55 | ||
More Information
Set Membership: |
|

Mrpl9
Genomic Map