Assay Details
| Gene Symbol: LOC101698292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857532.2 | XP_004857589.1 | 15 - 16 | 1966 | 95 | |
| XM_021238053.1 | XP_021093712.1 | 16 - 17 | 2024 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101698292| Gene Symbol: LOC101698292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857532.2 | XP_004857589.1 | 15 - 16 | 1966 | 95 | |
| XM_021238053.1 | XP_021093712.1 | 16 - 17 | 2024 | 95 | ||