Assay Details
Gene Symbol: Fbxw4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866354.3 | XP_004866411.1 | 5 - 6 | 766 | 59 | |
XM_021245594.1 | XP_021101253.1 | 5 - 6 | 700 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
