Assay Details
| Gene Symbol: Chrnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872090.3 | XP_004872147.1 | 3 - 4 | 407 | 60 | |
| XM_021250077.1 | XP_021105736.1 | 3 - 4 | 375 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021250077.1 transcripts.
Set Membership: |
|

Chrnb4
Genomic Map