Assay Details
| Gene Symbol: Prr16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836153.2 | XP_004836210.1 | 1 - 2 | 215 | 65 | |
| XM_013077780.2 | XP_012933234.1 | 1 - 2 | 215 | 65 | ||
| XM_021238072.1 | XP_021093731.1 | 1 - 2 | 215 | 65 | ||
More Information
Set Membership: |
|

Prr16
Genomic Map