Assay Details
| Gene Symbol: Pecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864620.3 | XP_004864677.1 | 5 - 6 | 678 | 69 | |
| XM_004864621.3 | XP_004864678.1 | 5 - 6 | 549 | 69 | ||
More Information
Set Membership: |
|

Pecr
Genomic Map