Assay Details
| Gene Symbol: LOC101721935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065925.2 | XP_012921379.1 | 2 - 3 | 249 | 84 | |
| XM_021244855.1 | XP_021100514.1 | 2 - 3 | 381 | 84 | ||
More Information
Set Membership: |
|

LOC101721935
Genomic Map