Assay Details
| Gene Symbol: Tsks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866883.2 | XP_004866940.1 | 7 - 8 | 1070 | 68 | |
| XM_004866884.2 | XP_004866941.1 | 7 - 8 | 1070 | 68 | ||
| XM_004866885.2 | XP_004866942.1 | 6 - 7 | 947 | 68 | ||
| XM_004866886.3 | XP_004866943.1 | 7 - 8 | 1070 | 68 | ||
| XM_004866888.2 | XP_004866945.1 | 6 - 7 | 947 | 68 | ||
| XM_004866889.2 | XP_004866946.1 | 7 - 8 | 1070 | 68 | ||
| XM_004866890.2 | XP_004866947.1 | 6 - 7 | 947 | 68 | ||
More Information
Set Membership: |
|

Tsks
Genomic Map