Assay Details
| Gene Symbol: LOC101713368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874080.3 | XP_004874137.1 | 4 - 5 | 440 | 75 | |
| XM_013069004.2 | XP_012924458.1 | 4 - 5 | 440 | 75 | ||
| XM_013069005.2 | XP_012924459.1 | 4 - 5 | 440 | 75 | ||
| XM_013069008.2 | XP_012924462.1 | 4 - 5 | 440 | 75 | ||
| XM_021251639.1 | XP_021107298.1 | 4 - 5 | 440 | 75 | ||
More Information
Set Membership: |
|

LOC101713368
Genomic Map