Assay Details
| Gene Symbol: Emilin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265195.1 | XP_021120854.1 | 7 - 8 | 2233 | 68 | |
| XM_021265196.1 | XP_021120855.1 | 6 - 7 | 1890 | 68 | ||
More Information
Set Membership: |
|

Emilin2
Genomic Map