Assay Details
| Gene Symbol: Neurl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866484.3 | XP_004866541.1 | 4 - 5 | 1391 | 53 | |
| XM_004866485.3 | XP_004866542.1 | 4 - 5 | 1505 | 53 | ||
| XM_021245693.1 | XP_021101352.1 | 4 - 5 | 1363 | 53 | ||
More Information
Set Membership: |
|

Neurl1
Genomic Map