Assay Details
| Gene Symbol: Slamf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239830.1 | XP_021095489.1 | 4 - 5 | 1718 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slamf9| Gene Symbol: Slamf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239830.1 | XP_021095489.1 | 4 - 5 | 1718 | 71 | |
Set Membership: |
|