Assay Details
| Gene Symbol: Mrps33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851289.3 | XP_004851346.1 | 2 - 3 | 364 | 70 | |
| XM_004851290.3 | XP_004851347.1 | 2 - 3 | 394 | 70 | ||
More Information
Set Membership: |
|

Mrps33
Genomic Map