Assay Details
| Gene Symbol: Chst13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854144.3 | XP_004854201.1 | 2 - 3 | 594 | 92 | |
| XM_021265991.1 | XP_021121650.1 | 1 - 2 | 686 | 92 | ||
More Information
Set Membership: |
|

Chst13
Genomic Map