Assay Details
| Gene Symbol: Znf213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864726.3 | XP_004864783.2 | 5 - 6 | 1156 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf213| Gene Symbol: Znf213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864726.3 | XP_004864783.2 | 5 - 6 | 1156 | 71 | |
Set Membership: |
|