Assay Details
| Gene Symbol: Znf771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856160.2 | XP_004856217.2 | 2 - 3 | 291 | 153 | |
| XM_004856161.3 | XP_004856218.1 | 2 - 3 | 366 | 153 | ||
| XM_021237457.1 | XP_021093116.1 | 2 - 3 | 236 | 153 | ||
More Information
Set Membership: |
|

Znf771
Genomic Map