Assay Details
| Gene Symbol: Akap12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856878.2 | XP_004856935.1 | 1 - 2 | 322 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap12| Gene Symbol: Akap12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856878.2 | XP_004856935.1 | 1 - 2 | 322 | 66 | |
Set Membership: |
|