Assay Details
Gene Symbol: Fam155a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858563.3 | XP_004858620.1 | 1 - 2 | 1446 | 65 | |
XM_021239416.1 | XP_021095075.1 | 1 - 2 | 1446 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
