Assay Details
| Gene Symbol: Wnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865257.2 | XP_004865314.1 | 2 - 3 | 547 | 65 | |
| XM_021244770.1 | XP_021100429.1 | 2 - 3 | 547 | 65 | ||
More Information
Set Membership: |
|

Wnt2
Genomic Map