Assay Details
| Gene Symbol: Fam83g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868764.2 | XP_004868821.1 | 3 - 4 | 1619 | 75 | |
| XM_004868765.3 | XP_004868822.1 | 3 - 4 | 1877 | 75 | ||
| XM_013067043.2 | XP_012922497.1 | 3 - 4 | 1876 | 75 | ||

Fam83g
Genomic Map