Assay Details
| Gene Symbol: Fam83g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868764.2 | XP_004868821.1 | 4 - 5 | 1746 | 75 | |
| XM_004868765.3 | XP_004868822.1 | 4 - 5 | 2004 | 75 | ||
| XM_013067043.2 | XP_012922497.1 | 4 - 5 | 2003 | 75 | ||
More Information
Set Membership: |
|

Fam83g
Genomic Map