Assay Details
| Gene Symbol: Fam20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870287.3 | XP_004870344.1 | 4 - 5 | 1419 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam20a| Gene Symbol: Fam20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870287.3 | XP_004870344.1 | 4 - 5 | 1419 | 53 | |
Set Membership: |
|