Assay Details
| Gene Symbol: Lyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872415.3 | XP_004872472.1 | 2 - 3 | 456 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyl1| Gene Symbol: Lyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872415.3 | XP_004872472.1 | 2 - 3 | 456 | 64 | |
Set Membership: |
|