Assay Details
| Gene Symbol: Pdzrn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874005.2 | XP_004874062.1 | 4 - 5 | 1082 | 64 | |
| XM_004874006.2 | XP_004874063.1 | 2 - 3 | 526 | 64 | ||
| XM_021251552.1 | XP_021107211.1 | 4 - 5 | 1219 | 64 | ||
More Information
Set Membership: |
|

Pdzrn4
Genomic Map