Assay Details
| Gene Symbol: LOC101723601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263412.1 | XP_021119071.1 | 1 - 2 | 37 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101723601| Gene Symbol: LOC101723601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263412.1 | XP_021119071.1 | 1 - 2 | 37 | 68 | |
Set Membership: |
|