Assay Details
| Gene Symbol: Chrac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837896.3 | XP_004837953.1 | 1 - 2 | 411 | 97 | |
| XM_004837897.3 | XP_004837954.1 | 1 - 2 | 402 | 97 | ||
| XM_004837898.3 | XP_004837955.1 | 1 - 2 | 406 | 97 | ||
| XM_004837899.3 | XP_004837956.1 | 1 - 2 | 407 | 97 | ||
More Information
Set Membership: |
|

Chrac1
Genomic Map