Assay Details
| Gene Symbol: Creld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845360.2 | XP_004845417.1 | 7 - 8 | 882 | 61 | |
| XM_004845361.2 | XP_004845418.1 | 7 - 8 | 882 | 61 | ||
More Information
Set Membership: |
|

Creld2
Genomic Map