Assay Details
| Gene Symbol: CUNH1orf198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854673.3 | XP_004854730.1 | 2 - 3 | 445 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH1orf198| Gene Symbol: CUNH1orf198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854673.3 | XP_004854730.1 | 2 - 3 | 445 | 58 | |
Set Membership: |
|