Assay Details
| Gene Symbol: Hs3st2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855935.2 | XP_004855992.1 | 1 - 2 | 604 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hs3st2| Gene Symbol: Hs3st2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855935.2 | XP_004855992.1 | 1 - 2 | 604 | 95 | |
Set Membership: |
|