Assay Details
| Gene Symbol: Sc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870913.3 | XP_004870970.1 | 4 - 5 | 621 | 89 | |
| XM_004870915.2 | XP_004870972.1 | 4 - 5 | 551 | 89 | ||
More Information
Set Membership: |
|

Sc5d
Genomic Map