Assay Details
| Gene Symbol: Thsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855543.2 | XP_004855600.1 | 5 - 6 | 972 | 56 | |
| XM_021236985.1 | XP_021092644.1 | 12 - 13 | 2124 | 56 | ||
More Information
Set Membership: |
|

Thsd4
Genomic Map