Assay Details
| Gene Symbol: Sept9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860792.2 | XP_004860849.1 | 5 - 6 | 1118 | 60 | |
| XM_004860793.3 | XP_004860850.1 | 4 - 5 | 1112 | 60 | ||
| XM_004860795.2 | XP_004860852.1 | 4 - 5 | 1049 | 60 | ||
| XM_004860798.3 | XP_004860855.1 | 3 - 4 | 735 | 60 | ||
| XM_021241257.1 | XP_021096916.1 | 4 - 5 | 1728 | 60 | ||
| XM_021241258.1 | XP_021096917.1 | 3 - 4 | 404 | 60 | ||
More Information
Set Membership: |
|

Sept9
Genomic Map