Assay Details
| Gene Symbol: Cpeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073101.2 | XP_012928555.1 | 12 - 13 | 1421 | 62 | |
| XM_013073108.2 | XP_012928562.1 | 10 - 11 | 1078 | 62 | ||
| XM_013073109.2 | XP_012928563.1 | 10 - 11 | 1252 | 62 | ||
| XM_013073110.2 | XP_012928564.1 | 9 - 10 | 1162 | 62 | ||
| XM_021260266.1 | XP_021115925.1 | 12 - 13 | 1672 | 62 | ||
| XM_021260267.1 | XP_021115926.1 | 11 - 12 | 1603 | 62 | ||
| XM_021260268.1 | XP_021115927.1 | 11 - 12 | 1582 | 62 | ||
| XM_021260269.1 | XP_021115928.1 | 10 - 11 | 1579 | 62 | ||
| XM_021260270.1 | XP_021115929.1 | 12 - 13 | 1570 | 62 | ||
| XM_021260271.1 | XP_021115930.1 | 10 - 11 | 1513 | 62 | ||
| XM_021260272.1 | XP_021115931.1 | 9 - 10 | 1489 | 62 | ||
| XM_021260273.1 | XP_021115932.1 | 12 - 13 | 1345 | 62 | ||
More Information
Set Membership: |
|

Cpeb2
Genomic Map