Assay Details
| Gene Symbol: Znf527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239457.1 | XP_021095116.1 | 1 - 2 | 915 | 100 | |
| XM_021239458.1 | XP_021095117.1 | 2 - 3 | 240 | 100 | ||
| XM_021239459.1 | XP_021095118.1 | 1 - 2 | 915 | 100 | ||
| XM_021239460.1 | XP_021095119.1 | 1 - 2 | 915 | 100 | ||
| XM_021239461.1 | XP_021095120.1 | 1 - 2 | 915 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021239458.1, XM_021239460.1, XM_021239461.1 transcripts.
Set Membership: |
|

Znf527
Genomic Map