Assay Details
| Gene Symbol: Tnfaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856975.3 | XP_004857032.1 | 2 - 3 | 545 | 62 | |
| XM_004856976.3 | XP_004857033.1 | 3 - 4 | 636 | 62 | ||
More Information
Set Membership: |
|

Tnfaip1
Genomic Map