Assay Details
| Gene Symbol: Zkscan8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847826.3 | XP_004847883.1 | 6 - 7 | 1229 | 102 | |
| XM_021260797.1 | XP_021116456.1 | 7 - 8 | 1334 | 102 | ||
| XM_021260799.1 | XP_021116458.1 | 7 - 8 | 1326 | 102 | ||
| XM_021260800.1 | XP_021116459.1 | 6 - 7 | 1224 | 102 | ||
| XM_021260801.1 | XP_021116460.1 | 6 - 7 | 1227 | 102 | ||
| XM_021260802.1 | XP_021116461.1 | 7 - 8 | 1214 | 102 | ||
| XM_021260803.1 | XP_021116462.1 | 6 - 7 | 1126 | 102 | ||
More Information
Set Membership: |
|

Zkscan8
Genomic Map