Assay Details
| Gene Symbol: Slc26a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854405.3 | XP_004854462.1 | 1 - 2 | 268 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004854405.3 transcripts.
Set Membership: |
|

Slc26a2
Genomic Map