Assay Details
| Gene Symbol: Slc26a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854405.3 | XP_004854462.1 | 2 - 3 | 984 | 73 | |
| XM_021266202.1 | XP_021121861.1 | 3 - 4 | 1152 | 73 | ||
| XM_021266203.1 | XP_021121862.1 | 3 - 4 | 1127 | 73 | ||
More Information
Set Membership: |
|

Slc26a2
Genomic Map